CAS RN 35896-58-3
CAS RN 35896-58-3
IUPAC Name:
1,2,3,4-tetramethoxy-5-methylbenzene
Synonyms:
1,2,3,4-tetramethoxy-5-methylbenzene
Molecular Weight (g/mol):
212.25
Molecular Formula:
C11H16O4
InChi Key:
OIWAVVSMXFIBCD-UHFFFAOYSA-N
SMILES:
COC1=C(OC)C(OC)=C(OC)C(C)=C1
1
–
1
of
1
results
1
2,3,4,5-Tetramethoxytoluene 97.0+%, TCI America™
CAS: 35896-58-3 Molecular Formula: C11H16O4 Molecular Weight (g/mol): 212.245 MDL Number: MFCD07370118 InChI Key: OIWAVVSMXFIBCD-UHFFFAOYSA-N PubChem CID: 9877764 IUPAC Name: 1,2,3,4-tetramethoxy-5-methylbenzene SMILES: CC1=CC(=C(C(=C1OC)OC)OC)OC