accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35897-34-8

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-carbamimidamidopentanoic acid hydrochloride
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-5-carbamimidamidopentanoic acid hydrochloride
Molecular Weight (g/mol): 310.78
Molecular Formula: C11H23ClN4O4
InChi Key: HDELGKMVZYHPPB-RZUXQYDTNA-N
SMILES: Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)