accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35899-43-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[1-(4-methylbenzenesulfonyl)-1H-imidazol-4-yl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[1-(4-methylbenzenesulfonyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 409.46
Molecular Formula: C18H23N3O6S
InChi Key: DCLJSEPKYJSEHW-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results