accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 359-41-1

IUPAC Name: (2S)-2-(trifluoromethyl)oxirane
Synonyms: (2S)-2-(trifluoromethyl)oxirane
Molecular Weight (g/mol): 112.05
Molecular Formula: C3H3F3O
InChi Key: AQZRARFZZMGLHL-REOHCLBHSA-N
SMILES: FC(F)(F)[C@@H]1CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results