accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOHOOOPOHOOPOHOOPOHOHONNONNH

CAS RN 35908-31-7

IUPAC Name: sodium ({[({[3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid
Synonyms: sodium ({[3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy(hydroxy)phosphoryl)oxyphosphonic acid
Molecular Weight (g/mol): 531.16
Molecular Formula: C10H15N4NaO14P3
InChi Key: AUOLCGMICMHAHB-UHFFFAOYNA-N
SMILES: [Na+].OC1C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)OC(C1O)N1C=NC2=C1NC=NC2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results