accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3592-12-9

IUPAC Name: 5,5-dimethyl-1,3-dioxan-2-one
Synonyms: 5,5-dimethyl-1,3-dioxan-2-one
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: JRFXQKZEGILCCO-UHFFFAOYSA-N
SMILES: CC1(C)COC(=O)OC1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results