accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNCSFF

CAS RN 35923-79-6

IUPAC Name: 1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene
Synonyms: 1,2,3,4,5-pentafluoro-6-isothiocyanatobenzene
Molecular Weight (g/mol): 225.14
Molecular Formula: C7F5NS
InChi Key: NGNKMRBGZPDABE-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(N=C=S)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results