accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35944-64-0

IUPAC Name: 3-iodo-4-methylaniline
Synonyms: 3-iodo-4-methylaniline
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: RRUDMHNAMZFNEK-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1I
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)