accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 3598-14-9

IUPAC Name: 2-phenoxyacetonitrile
Synonyms: 2-phenoxyacetonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: VLLSCJFPVSQXDM-UHFFFAOYSA-N
SMILES: N#CCOC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results