accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 359860-27-8

IUPAC Name: N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-(2-{2-[2-(2-aminoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 418.55
Molecular Formula: C18H34N4O5S
InChi Key: KIJSBKNJFAUJFV-UHFFFAOYNA-N
SMILES: NCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results