accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35999-20-3

IUPAC Name: 3-chloro-1-(4-methoxyphenyl)propan-1-one
Synonyms: 3-chloro-1-(4-methoxyphenyl)propan-1-one
Molecular Weight (g/mol): 198.65
Molecular Formula: C10H11ClO2
InChi Key: FJBUZSAECLLZOL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)CCCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results