accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3601-36-3

IUPAC Name: [4,5-bis(benzoyloxy)-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
Synonyms: [4,5-bis(benzoyloxy)-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
Molecular Weight (g/mol): 506.51
Molecular Formula: C28H26O9
InChi Key: WXFFEILSURAFKL-UHFFFAOYNA-N
SMILES: COC1OC(COC(=O)C2=CC=CC=C2)C(O)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)