accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHOOHNN

CAS RN 36023-64-0

IUPAC Name: 5,6-dioxo-1,4,5,6-tetrahydropyrazine-2,3-dicarbonitrile
Synonyms: 5,6-dioxo-1,4-dihydropyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 162.11
Molecular Formula: C6H2N4O2
InChi Key: OTVDGBINMCDFTF-UHFFFAOYSA-N
SMILES: O=C1NC(C#N)=C(NC1=O)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results