CAS RN 360576-04-1
IUPAC Name:
6-bromo-2,3-difluorobenzaldehyde
Synonyms:
6-bromo-2,3-difluorobenzaldehyde
Molecular Weight (g/mol):
221
Molecular Formula:
C7H3BrF2O
InChi Key:
LAVPYRPTHABUAD-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(C=O)=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
6-Bromo-2,3-difluorobenzaldehyde, 98%
6-Bromo-2,3-difluorobenzaldehyde, 98%
CAS: 360576-04-1 Molecular Formula: C7H3BrF2O Molecular Weight (g/mol): 221.001 MDL Number: MFCD07368119 InChI Key: LAVPYRPTHABUAD-UHFFFAOYSA-N Synonym: benzaldehyde, 6-bromo-2,3-difluoro,6-bromo-2,3-difluoro-benzaldehyde,2-bromo-5,6-difluorobenzaldehyde,benzaldehyde,6-bromo-2,3-difluoro PubChem CID: 9877851 IUPAC Name: 6-bromo-2,3-difluorobenzaldehyde SMILES: C1=CC(=C(C(=C1F)F)C=O)Br