accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36070-80-1

IUPAC Name: 5-chloropyrazine-2-carboxylic acid
Synonyms: 5-chloropyrazine-2-carboxylic acid
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: FXJOTWLLDJYKAG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(Cl)C=N1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)