CAS RN 360768-57-6
IUPAC Name:
4-ethynyl-4'-propyl-1,1'-biphenyl
Synonyms:
4-ethynyl-4'-propyl-1,1'-biphenyl
Molecular Weight (g/mol):
220.32
Molecular Formula:
C17H16
InChi Key:
ZBSZCHDBZRCUES-UHFFFAOYSA-N
SMILES:
CCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#C
1
–
1
of
1
results
1
4-Ethynyl-4'-propylbiphenyl 98.0+%, TCI America™
CAS: 360768-57-6 Molecular Formula: C17H16 Molecular Weight (g/mol): 220.315 MDL Number: MFCD18072535 InChI Key: ZBSZCHDBZRCUES-UHFFFAOYSA-N PubChem CID: 19814911 IUPAC Name: 1-ethynyl-4-(4-propylphenyl)benzene SMILES: CCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#C