accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36082-50-5

IUPAC Name: 5-bromo-2,4-dichloropyrimidine
Synonyms: 5-bromo-2,4-dichloropyrimidine
Molecular Weight (g/mol): 227.87
Molecular Formula: C4HBrCl2N2
InChi Key: SIKXIUWKPGWBBF-UHFFFAOYSA-N
SMILES: ClC1=NC=C(Br)C(Cl)=N1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)