accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHO

CAS RN 3612-18-8

IUPAC Name: 1-ethyl-4-oxopiperidin-1-ium
Synonyms: 1-ethyl-4-oxopiperidin-1-ium
Molecular Weight (g/mol): 128.19
Molecular Formula: C7H14NO
InChi Key: BDVKAMAALQXGLM-UHFFFAOYSA-O
SMILES: CC[NH+]1CCC(=O)CC1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)