accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFF

CAS RN 361393-85-3

IUPAC Name: 1-[4-(trifluoromethyl)cyclohexyl]methanamine
Synonyms: 1-[4-(trifluoromethyl)cyclohexyl]methanamine
Molecular Weight (g/mol): 181.2
Molecular Formula: C8H14F3N
InChi Key: XAKIXQQQAQHCMN-UHFFFAOYSA-N
SMILES: NCC1CCC(CC1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results