CAS RN 361442-04-8
IUPAC Name:
2-[2-amino-2-(3-hydroxyadamantan-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile
Synonyms:
saxagliptin
Molecular Weight (g/mol):
315.42
Molecular Formula:
C18H25N3O2
InChi Key:
QGJUIPDUBHWZPV-UHFFFAOYNA-N
SMILES:
NC(C(=O)N1C2CC2CC1C#N)C12CC3CC(CC(O)(C3)C1)C2
1
–
1
of
1
results
1
Promotions Available
Saxagliptin
Saxagliptin
CAS: 361442-04-8 Molecular Formula: C18H25N3O2 Molecular Weight (g/mol): 315.42 MDL Number: MFCD12756398 InChI Key: QGJUIPDUBHWZPV-UHFFFAOYNA-N Synonym: saxagliptin PubChem CID: 129317852 IUPAC Name: (1R,5R)-2-[2-amino-2-[(7R)-3-hydroxy-1-adamantyl]acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile SMILES: NC(C(=O)N1C2CC2CC1C#N)C12CC3CC(CC(O)(C3)C1)C2