accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNNH2

CAS RN 3619-22-5

IUPAC Name: 4-methylbenzohydrazide
Synonyms: benzoylhydrazine, P-methyl
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: MFFVZXOPRXMVET-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NN
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)