CAS RN 36193-65-4
CAS RN 36193-65-4
IUPAC Name:
1H-indole-2-carbonitrile
Synonyms:
1H-indole-2-carbonitrile
Molecular Weight (g/mol):
142.16
Molecular Formula:
C9H6N2
InChi Key:
CBTITARLOCZPDU-UHFFFAOYSA-N
SMILES:
N#CC1=CC2=CC=CC=C2N1
1
–
1
of
1
results
1
Indole-2-carbonitrile, 97%
CAS: 36193-65-4 Molecular Formula: C9H6N2 Molecular Weight (g/mol): 142.16 MDL Number: MFCD00187566 InChI Key: CBTITARLOCZPDU-UHFFFAOYSA-N Synonym: 2-cyanoindole,cyanoindole,2-indolecarbonitrile,2-cyano-1h-indole,indole-2-carbonitrile,racemic indole nitrile PubChem CID: 3787599 IUPAC Name: 1H-indole-2-carbonitrile SMILES: N#CC1=CC2=CC=CC=C2N1