accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36216-80-5

IUPAC Name: 1,2-benzoxazol-3-amine
Synonyms: 1,2-benzoxazol-3-amine
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: NLMVYUBGWZWUGB-UHFFFAOYSA-N
SMILES: NC1=NOC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results