accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHC

CAS RN 3623-15-2

IUPAC Name: 1-phenylprop-2-yn-1-one
Synonyms: 1-phenylprop-2-yn-1-one
Molecular Weight (g/mol): 130.15
Molecular Formula: C9H6O
InChi Key: JITPLZPWKYUTDM-UHFFFAOYSA-N
SMILES: O=C(C#C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)