accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3NNH2O

CAS RN 3625-57-8

IUPAC Name: [9-(diethylamino)benzo[a]phenoxazin-5-ylidene]azanium;sulfate
Synonyms: nile blue cresyl fast violet cresol fast violet nile blue dye nile blue chloride
Molecular Weight (g/mol): 732.84
Molecular Formula: C40H40N6O6S
InChi Key: QIRDPEPUXNCOLD-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.CCN(CC)C1=CC2=C(C=C1)N=C3C4=CC=CC=C4C(=[NH2+])C=C3O2.[O-]S(=O)(=O)[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)