accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 362670-07-3

IUPAC Name: tert-butyl N-(2-amino-5-fluorophenyl)carbamate
Synonyms: tert-butyl N-(2-amino-5-fluorophenyl)carbamate
Molecular Weight (g/mol): 226.25
Molecular Formula: C11H15FN2O2
InChi Key: FELBQUPQCJZQQX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=C(N)C=CC(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results