accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBr

CAS RN 36276-24-1

IUPAC Name: 3-bromo-4-methylbenzaldehyde
Synonyms: 3-bromo-4-methylbenzaldehyde
Molecular Weight (g/mol): 199.05
Molecular Formula: C8H7BrO
InChi Key: WTXXUAHMTVAQHW-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)