accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36282-40-3

IUPAC Name: bromo(3-methoxyphenyl)magnesium
Synonyms: bromo(3-methoxyphenyl)magnesium
Molecular Weight (g/mol): 211.34
Molecular Formula: C7H7BrMgO
InChi Key: AJKNYEHIJAMGHX-UHFFFAOYSA-M
SMILES: COC1=CC=CC([Mg]Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)