accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 363-58-6

IUPAC Name: ethyl (2S)-2-chloro-4,4,4-trifluoro-3-oxobutanoate
Synonyms: ethyl (2S)-2-chloro-4,4,4-trifluoro-3-oxobutanoate
Molecular Weight (g/mol): 218.56
Molecular Formula: C6H6ClF3O3
InChi Key: YVWUNJVPOCYLIM-VKHMYHEASA-N
SMILES: CCOC(=O)[C@@H](Cl)C(=O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results