accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFF

CAS RN 363-72-4

IUPAC Name: 1,2,3,4,5-pentafluorobenzene
Synonyms: pentafluorobenzene
Molecular Weight (g/mol): 168.07
Molecular Formula: C6HF5
InChi Key: WACNXHCZHTVBJM-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (6)
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)