accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36309-88-3

IUPAC Name: tris(4,7-diphenyl-1,10-phenanthroline); dichlororuthenium
Synonyms: tris(4,7-diphenyl-1,10-phenanthroline); dichlororuthenium
Molecular Weight (g/mol): 1169.19
Molecular Formula: C72H48Cl2N6Ru
InChi Key: SKZWFYFFTOHWQP-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.C1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results