CAS RN 36321-73-0
IUPAC Name:
1,2-dibromo-3,4,5,6-tetramethylbenzene
Synonyms:
1,2-dibromo-3,4,5,6-tetramethylbenzene
Molecular Weight (g/mol):
292.01
Molecular Formula:
C10H12Br2
InChi Key:
VEZJOZSIGSNYEB-UHFFFAOYSA-N
SMILES:
CC1=C(C)C(C)=C(Br)C(Br)=C1C
1
–
1
of
1
results
1
Promotions Available
1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%
1,2-Dibromo-3,4,5,6-tetramethylbenzene, 98%
CAS: 36321-73-0 Molecular Formula: C10H12Br2 Molecular Weight (g/mol): 292.014 MDL Number: MFCD00154987 InChI Key: VEZJOZSIGSNYEB-UHFFFAOYSA-N Synonym: benzene,1,2-dibromo-3,4,5,6-tetramethyl,pubchem21478,benzene, 1,2-dibromo-3,4,5,6-tetramethyl,1,2-bis bromanyl-3,4,5,6-tetramethyl-benzene PubChem CID: 2752606 IUPAC Name: 1,2-dibromo-3,4,5,6-tetramethylbenzene SMILES: CC1=C(C(=C(C(=C1C)Br)Br)C)C