CAS RN 36323-28-1
IUPAC Name:
1,3-bis(dibromomethyl)benzene
Synonyms:
1,3-bis(dibromomethyl)benzene
Molecular Weight (g/mol):
421.75
Molecular Formula:
C8H6Br4
InChi Key:
ZMCUKNMLHBAGMS-UHFFFAOYSA-N
SMILES:
BrC(Br)C1=CC(=CC=C1)C(Br)Br
1
–
1
of
1
results
1
alpha,alpha,alpha',alpha'-Tetrabromo-m-xylene 96.0+%, TCI America™
CAS: 36323-28-1 Molecular Formula: C8H6Br4 Molecular Weight (g/mol): 421.752 MDL Number: MFCD00000132 InChI Key: ZMCUKNMLHBAGMS-UHFFFAOYSA-N Synonym: 1,3-Bis(dibromomethyl)benzene, m-Xylylene Tetrabromide PubChem CID: 37418 IUPAC Name: 1,3-bis(dibromomethyl)benzene SMILES: C1=CC(=CC(=C1)C(Br)Br)C(Br)Br