accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3H3C

CAS RN 3637-01-2

IUPAC Name: 1-(3,4-dimethylphenyl)ethan-1-one
Synonyms: 1-(3,4-dimethylphenyl)ethanone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: WPRAXAOJIODQJR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C)C(C)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results