accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36393-42-7

IUPAC Name: 4'-methyl-[1,1'-biphenyl]-4-carbaldehyde
Synonyms: 4'-methyl-[1,1'-biphenyl]-4-carbaldehyde
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: BCINBWXQYBLSKO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results