accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36394-75-9

IUPAC Name: 1-chloro-1-oxopropan-2-yl acetate
Synonyms: 1-chloro-1-oxopropan-2-yl acetate
Molecular Weight (g/mol): 150.56
Molecular Formula: C5H7ClO3
InChi Key: ALHZEIINTQJLOT-UHFFFAOYNA-N
SMILES: CC(OC(C)=O)C(Cl)=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)