accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFNOONOO

CAS RN 364-32-9

IUPAC Name: 4-fluoro-2,6-dinitrophenol
Synonyms: 4-fluoro-2,6-dinitrophenol
Molecular Weight (g/mol): 202.1
Molecular Formula: C6H3FN2O5
InChi Key: MDOWEUXXLVBZIU-UHFFFAOYSA-N
SMILES: OC1=C(C=C(F)C=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results