accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 364-76-1

IUPAC Name: 4-fluoro-3-nitroaniline
Synonyms: 4-fluoro-3-nitroaniline
Molecular Weight (g/mol): 156.12
Molecular Formula: C6H5FN2O2
InChi Key: LLIOADBCFIXIEU-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (7)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (6)
  • (1)
  • (7)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)