accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 364-77-2

ONOFI

CAS RN 364-77-2

IUPAC Name: 4-fluoro-1-iodo-2-nitrobenzene
Synonyms: 4-fluoro-1-iodo-2-nitrobenzene
Molecular Weight (g/mol): 267
Molecular Formula: C6H3FINO2
InChi Key: ANPFMDLUEWNKIM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results