accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNO

CAS RN 36404-88-3

IUPAC Name: 2-chloropyridine-3-carbaldehyde
Synonyms: 2-chloropyridine-3-carbaldehyde
Molecular Weight (g/mol): 141.55
Molecular Formula: C6H4ClNO
InChi Key: KHPAGGHFIDLUMB-UHFFFAOYSA-N
SMILES: ClC1=NC=CC=C1C=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (6)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)