accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrH3CNNCH3

CAS RN 36437-30-6

IUPAC Name: 1,1'-dioctyl-[4,4'-bipyridine]-1,1'-diium dibromide
Synonyms: 1,1'-dioctyl-[4,4'-bipyridine]-1,1'-diium dibromide
Molecular Weight (g/mol): 542.44
Molecular Formula: C26H42Br2N2
InChi Key: GVUBEJANEHZYPA-UHFFFAOYSA-L
SMILES: [Br-].[Br-].CCCCCCCC[N+]1=CC=C(C=C1)C1=CC=[N+](CCCCCCCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results