accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36443-80-8

IUPAC Name: 1-benzyl-3-methyl-1H-imidazol-3-ium chloride
Synonyms: 3-benzyl-1-methylimidazol-1-ium chloride
Molecular Weight (g/mol): 208.69
Molecular Formula: C11H13ClN2
InChi Key: FCRZSGZCOGHOGF-UHFFFAOYSA-M
SMILES: [Cl-].C[N+]1=CN(CC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)