CAS RN 3645-00-9
CAS RN 3645-00-9
IUPAC Name:
1-(2-hydroxyethyl) 4-methyl benzene-1,4-dicarboxylate
Synonyms:
1-(2-hydroxyethyl) 4-methyl benzene-1,4-dicarboxylate
Molecular Weight (g/mol):
224.21
Molecular Formula:
C11H12O5
InChi Key:
DJQMYWWZWUOCBQ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(C=C1)C(=O)OCCO
1
–
2
of
2
results
1
2-Hydroxyethyl Methyl Terephthalate 97.0+%, TCI America™
CAS: 3645-00-9 Molecular Formula: C11H12O5 Molecular Weight (g/mol): 224.212 MDL Number: MFCD01321191 InChI Key: DJQMYWWZWUOCBQ-UHFFFAOYSA-N Synonym: Terephthalic Acid 2-Hydroxyethyl Methyl Ester PubChem CID: 77207 IUPAC Name: 4-O-(2-hydroxyethyl) 1-O-methyl benzene-1,4-dicarboxylate SMILES: COC(=O)C1=CC=C(C=C1)C(=O)OCCO
2
Sigma Aldrich 1-(2-Hydroxyethyl) 4-methyl terephthalate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.