accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3645-34-9

IUPAC Name: titanium(4+) tetrakis(2-ethylhexanoate)
Synonyms: titanium(4+) tetrakis(2-ethylhexanoate)
Molecular Weight (g/mol): 620.69
Molecular Formula: C32H60O8Ti
InChi Key: GDKAXSGMPFSRJY-UHFFFAOYNA-J
SMILES: [Ti+4].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results