accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNNOSClH2OHClHCl

CAS RN 364590-63-6

IUPAC Name: 3-chloro-N-[4-(methylamino)cyclohexyl]-N-{[3-(pyridin-4-yl)phenyl]methyl}-1-benzothiophene-2-carboxamide hydrate dihydrochloride
Synonyms: smoothened agonist hydrate dihydrochloride
Molecular Weight (g/mol): 580.99
Molecular Formula: C28H32Cl3N3O2S
InChi Key: IYXUQUYRWPGIQL-UHFFFAOYSA-N
SMILES: O.Cl.Cl.CNC1CCC(CC1)N(CC1=CC(=CC=C1)C1=CC=NC=C1)C(=O)C1=C(Cl)C2=CC=CC=C2S1
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results