accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 364622-82-2

IUPAC Name: (4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-3-{[(3S,5S)-5-[(sulfamoylamino)methyl]pyrrolidin-3-yl]sulfanyl}-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate
Synonyms: doripenem hydrate
Molecular Weight (g/mol): 438.51
Molecular Formula: C15H26N4O7S2
InChi Key: NTUBEBXBDGKBTJ-WGLOMNHJSA-N
SMILES: O.C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@@H]3CN[C@H](CNS(N)(=O)=O)C3)=C(N2C1=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results