accessibility menu, dialog, popup

UserName

Viewing profile as user:
PP

CAS RN 364732-88-7

IUPAC Name: [11-(diphenylphosphanyl)tricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaen-5-yl]diphenylphosphane
Synonyms: phanephos
Molecular Weight (g/mol): 576.66
Molecular Formula: C40H34P2
InChi Key: GYZZZILPVUYAFJ-UHFFFAOYSA-N
SMILES: C1CC2=C(C=C(CCC3=C(C=C1C=C3)P(C1=CC=CC=C1)C1=CC=CC=C1)C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results