accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONSBr

CAS RN 364793-76-0

IUPAC Name: 4-[(5-bromothiophen-2-yl)methyl]morpholine
Synonyms: 4-[(5-bromothiophen-2-yl)methyl]morpholine
Molecular Weight (g/mol): 262.17
Molecular Formula: C9H12BrNOS
InChi Key: XTTAOCHKCUOJFW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CN2CCOCC2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results