accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3648-21-3

IUPAC Name: 1,2-diheptyl benzene-1,2-dicarboxylate
Synonyms: DHPP di-n-heptylphthalate N-diheptyl phthalate
Molecular Weight (g/mol): 362.51
Molecular Formula: C22H34O4
InChi Key: JQCXWCOOWVGKMT-UHFFFAOYSA-N
SMILES: CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC
Percent Purity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)