accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36480-40-7

HSClCl

CAS RN 36480-40-7

IUPAC Name: (3,4-dichlorophenyl)methanethiol
Synonyms: (3,4-dichlorophenyl)methanethiol
Molecular Weight (g/mol): 193.09
Molecular Formula: C7H6Cl2S
InChi Key: CEGBRSQPRQXALB-UHFFFAOYSA-N
SMILES: SCC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)